3,5-Bis(trifluormethyl)benzoesäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC3090-100G
PC3090-10G
PC3090-5G
PC3090-25G
APOSPC3090-100GEA
87
EUR
APOSPC3090-100G
APOSPC3090-10G
APOSPC3090-5G
APOSPC3090-25G
3,5-Bis(trifluormethyl)benzoesäure 97%
3,5-Bis(trifluormethyl)benzoesäure
Formel:
C₉H₄F₆O₂ Molecular Weight: 258,12 g/mol Schmelzpunkt: 140…144 °C Dichte: 1,42 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000388 CAS-Nummer: 725-89-3 EINECS: 211-970-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...