3,5-Bis(trifluormethyl)benzhydrazid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Bis(trifluoromethyl)benzohydrazide

PC3090C-100G PC3090C-25G
APOSPC3090C-100GEA 401 EUR
APOSPC3090C-100G APOSPC3090C-25G
3,5-Bis(trifluormethyl)benzhydrazid 97%
3,5-Bis(trifluormethyl)benzhydrazid
Formel: C₉H₆F₆N₂O
Molecular Weight: 272,15 g/mol
Schmelzpunkt: 132…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051848
CAS-Nummer: 26107-82-4
EINECS: 247-462-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR