3,4'-Bis(trifluormethyl)benzophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(Trifluormethyl)-3-((4-(trifluormethyl)phenyl)methanoyl)benzene
PC3102-25G
PC3102-5G
PC3102-10G
APOSPC3102-25GEA
366
EUR
APOSPC3102-25G
APOSPC3102-5G
APOSPC3102-10G
3,4'-Bis(trifluormethyl)benzophenon 98%
3,4'-Bis(trifluormethyl)benzophenon
Formel:
C₁₅H₈F₆O Molecular Weight: 318.22 g/mol Schmelzpunkt: 93…97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013561 CAS-Nummer: 21084-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...