2,4-Bis(trifluormethyl)benzoylchlorid 98%

Lieferant: Apollo Scientific
Gefahr

PC3103D-1G PC3103D-25G PC3103D-100G
APOSPC3103D-1GEA 21.9 EUR
APOSPC3103D-1G APOSPC3103D-25G APOSPC3103D-100G
2,4-Bis(trifluormethyl)benzoylchlorid 98%
2,4-Bis(trifluormethyl)benzoylchlorid
Formel: C₉H₃ClF₆O
Molecular Weight: 276,57 g/mol
Siedepunkt: 179 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061227
CAS-Nummer: 53130-43-1
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR