1H,1H,7H-Perfluoroheptyl methacrylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Methacrylate, Methacrylic acid 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl ester

PC3132F-5G PC3132F-25G
APOSPC3132F-5GEA 72.5 EUR
APOSPC3132F-5G APOSPC3132F-25G
1H,1H,7H-Perfluoroheptyl methacrylate 97%
1H,1H,7H-Perfluoroheptyl methacrylate

Prone to polymerisation, may require distillation if not used within 3 months

Formel: C₁₁H₈F₁₂O₂
Molecular Weight: 400,17 g/mol
Siedepunkt: 112 °C/40mmHg
Dichte: 1,536 g/cm³ (25 °C)
Flammpunkt: 104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00080658
CAS-Nummer: 2261-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR