4-(Trifluoromethyl)phenyl acetate 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC31334-1G
PC31334-5G
APOSPC31334-1GEA
241
EUR
APOSPC31334-1G
APOSPC31334-5G
4-(Trifluoromethyl)phenyl acetate 95%
4-(Trifluoromethyl)phenyl acetate
Formel:
C₉H₇F₃O₂ Flammpunkt: 69 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00829056 CAS-Nummer: 78950-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...