6-Fluoro-4H-benzo[1,3]dioxine-8-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC31428-1G
PC31428-10G
APOSPC31428-1GEA
136
EUR
APOSPC31428-1G
APOSPC31428-10G
6-Fluoro-4H-benzo[1,3]dioxine-8-carboxylic acid
6-Fluoro-4H-benzo[1,3]dioxine-8-carboxylic acid
Formel:
C₉H₇FO₄ Schmelzpunkt: 180…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02681981 CAS-Nummer: 321309-28-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...