1-[2-Nitro-4-(trifluoromethyl)phenyl]-1H-pyrrole

Lieferant: Apollo Scientific

PC31460-1G PC31460-250MG PC31460-500MG PC31460-2.5G
APOSPC31460-1GEA 184 EUR
APOSPC31460-1G APOSPC31460-250MG APOSPC31460-500MG APOSPC31460-2.5G
1-[2-Nitro-4-(trifluoromethyl)phenyl]-1H-pyrrole
1-[2-Nitro-4-(trifluoromethyl)phenyl]-1H-pyrrole
Formel: C₁₁H₇F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00119450
CAS-Nummer: 136773-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR