2-Chlor-6-fluor-3-nitrobenzonitril
Lieferant: Apollo Scientific
|
Gefahr
|
PC31540-1G
PC31540-5G
APOSPC31540-1GEA
125
EUR
APOSPC31540-1G
APOSPC31540-5G
2-Chlor-6-fluor-3-nitrobenzonitril
2-Chlor-6-fluor-3-nitrobenzonitril
Formel:
C₇H₂ClFN₂O₂ Schmelzpunkt: 91…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053834 CAS-Nummer: 226419-18-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...