Ethylbromdifluoracetat 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Bromdifluoressigsäureethylester

PC3162-50G PC3162-250G PC3162-500G
APOSPC3162-50GEA 21.9 EUR
APOSPC3162-50G APOSPC3162-250G APOSPC3162-500G
Ethylbromdifluoracetat 98%
Ethylbromdifluoracetat

Treatment of the Reformatsky reagent with aldehydes and ketones affords 2,2-difluoro-3-hydroxy esters. Please enquire for references and other data for this item.

Formel: BrF₂CCOOCH₂CH₃
Molecular Weight: 202,98 g/mol
Siedepunkt: 112 °C/700mmHg
Dichte: 1,583 g/cm³ (25 °C)
Flammpunkt: 21 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042069
CAS-Nummer: 667-27-6
EINECS: 211-567-0
UN: 2924
ADR: 3+8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR