Ethyl-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
PC31682-5G
APOSPC31682-5GEA
77
EUR
APOSPC31682-5G
Ethyl-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
Ethyl-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
Formel:
C₈H₆ClF₃N₂O₂ Molecular Weight: 254,6 g/mol Dichte: 1,45 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01312372 CAS-Nummer: 187035-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...