Ethyl-6-chloro-5-cyano-2-(trifluoromethyl)nicotinate 99%

Lieferant: Apollo Scientific
Achtung

PC3169-1G PC3169-250MG PC3169-5G
APOSPC3169-1GEA 63.8 EUR
APOSPC3169-1G APOSPC3169-250MG APOSPC3169-5G
Ethyl-6-chloro-5-cyano-2-(trifluoromethyl)nicotinate 99%
Ethyl-6-chloro-5-cyano-2-(trifluoromethyl)nicotinate
Formel: C₁₀H₆ClF₃N₂O₂
Molecular Weight: 278.615 g/mol
Schmelzpunkt: 78…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00178758
CAS-Nummer: 175277-73-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR