Methyl-2-[4-(trifluoromethyl)piperidino]acetate
Lieferant: Apollo Scientific
PC3177-1G
PC3177-500MG
APOSPC3177-1GEA
336
EUR
APOSPC3177-1G
APOSPC3177-500MG
Methyl-2-[4-(trifluoromethyl)piperidino]acetate
Methyl-2-[4-(trifluoromethyl)piperidino]acetate
Formel:
C₉H₁₄F₃NO₂ Molecular Weight: 225,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09865019 CAS-Nummer: 860343-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...