1,1,2,3,3,3-Hexafluorpropyldifluormethylether
Lieferant: Apollo Scientific
|
Achtung
|
PC3178-5G
PC3178-25G
APOSPC3178-5GEA
0
EUR
APOSPC3178-5G
APOSPC3178-25G
1,1,2,3,3,3-Hexafluorpropyldifluormethylether
1,1,2,3,3,3-Hexafluorpropyldifluormethylether
Formel:
C₄H₂F₈O Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094220 CAS-Nummer: 56860-85-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...