2-Fluor-6-methoxypyridin
Lieferant: Apollo Scientific
|
Achtung
|
PC3181-5G
APOSPC3181-5GEA
21.9
EUR
APOSPC3181-5G
2-Fluor-6-methoxypyridin
2-Fluoro-6-methoxypyridine
Formel:
C₆H₆FNO Flammpunkt: <60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09998161 CAS-Nummer: 116241-61-3 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...