Diethyl (4-chloro-2-fluorophenyl)malonate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Diethyl 2-(4-chloro-2-fluorophenyl)malonate
PC31868-1G
PC31868-250MG
PC31868-5G
APOSPC31868-1GEA
249
EUR
APOSPC31868-1G
APOSPC31868-250MG
APOSPC31868-5G
Diethyl (4-chloro-2-fluorophenyl)malonate 95%
Diethyl (4-chloro-2-fluorophenyl)malonate
Formel:
C₁₃H₁₄ClFO₄ Molecular Weight: 288,71 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01570544 CAS-Nummer: 680211-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...