(2,4-Bis(trifluormethyl)phenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Bis(trifluoromethyl)phenylacetic acid

PC3202-1G PC3202-5G
APOSPC3202-1GEA 34.8 EUR
APOSPC3202-1G APOSPC3202-5G
(2,4-Bis(trifluormethyl)phenyl)essigsäure 98%
(2,4-Bis(trifluormethyl)phenyl)essigsäure
Formel: (C₆H₃(CF₃)₂)OCH₂COOH
Molecular Weight: 272,15 g/mol
Schmelzpunkt: 87,5…87.9 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042492
CAS-Nummer: 177952-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR