2'-Fluor-5'-nitroacetophenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Fluoro-5-nitrophenyl)ethanone
PC32093-25G
PC32093-5G
PC32093-100G
APOSPC32093-25GEA
59.6
EUR
APOSPC32093-25G
APOSPC32093-5G
APOSPC32093-100G
2'-Fluor-5'-nitroacetophenon
2'-Fluor-5'-nitroacetophenon
Formel:
C₈H₆FNO₃ Molecular Weight: 183,1400 g/mol Schmelzpunkt: 46…50 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00277867 CAS-Nummer: 79110-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...