1-(5-Amino-2-fluorophenyl)ethanone 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5'-Amino-2'-fluoroacetophenone

PC32127-1G PC32127-5G PC32127-10G PC32127-25G
APOSPC32127-1GEA 56.7 EUR
APOSPC32127-1G APOSPC32127-5G APOSPC32127-10G APOSPC32127-25G
1-(5-Amino-2-fluorophenyl)ethanone 95%
1-(5-Amino-2-fluorophenyl)ethanone
Formel: C₈H₈FNO
Molecular Weight: 153,16 g/mol
Schmelzpunkt: 68…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02090052
CAS-Nummer: 67500-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR