Ethyl {[4-(trifluoromethoxy)phenyl]amino}acetate
Lieferant: Apollo Scientific
|
Achtung
|
PC32272-1G
PC32272-250MG
PC32272-500MG
APOSPC32272-1GEA
105
EUR
APOSPC32272-1G
APOSPC32272-250MG
APOSPC32272-500MG
Ethyl {[4-(trifluoromethoxy)phenyl]amino}acetate
Ethyl {[4-(trifluoromethoxy)phenyl]amino}acetate
Formel:
C₁₁H₁₂F₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00176763 CAS-Nummer: 306935-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...