3-(4-Methoxyphenyl)-1,1,1-trifluoro-2-propanone
Lieferant: Apollo Scientific
Synonyme:
3-(4-Methoxyphenyl)-1,1,1-trifluoroacetone
PC32341-1G
PC32341-5G
APOSPC32341-1GEA
364
EUR
APOSPC32341-1G
APOSPC32341-5G
3-(4-Methoxyphenyl)-1,1,1-trifluoro-2-propanone
3-(4-Methoxyphenyl)-1,1,1-trifluoro-2-propanone
Formel:
C₁₀H₉F₃O₂ Molecular Weight: 218,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01764734 CAS-Nummer: 22102-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...