Ethyl-2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

PC3246-250MG PC3246-1G PC3246-5G
APOSPC3246-250MGEA 77 EUR
APOSPC3246-250MG APOSPC3246-1G APOSPC3246-5G
Ethyl-2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate 97%
Ethyl-2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate
Formel: C₈H₈F₃NO₂S
Molecular Weight: 239.22 g/mol
Schmelzpunkt: 34…37 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00153150
CAS-Nummer: 117724-62-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR