2-(Trifluormethylthio)phenylacetonitril 98%

Lieferant: Apollo Scientific
Achtung

PC3266-1G PC3266-5G
APOSPC3266-1GEA 143 EUR
APOSPC3266-1G APOSPC3266-5G
2-(Trifluormethylthio)phenylacetonitril 98%
2-(Trifluormethylthio)phenylacetonitril
Formel: C₉H₆F₃NS
Lagertemperatur: Raumtemperatur
MDL: MFCD00236352
CAS-Nummer: 237424-20-3
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR