6-Fluor-2-mercaptobenzothiazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Fluorobenzo[d]thiazole-2(3H)-thione

PC3267-1G PC3267-5G
APOSPC3267-1GEA 62.5 EUR
APOSPC3267-1G APOSPC3267-5G
6-Fluor-2-mercaptobenzothiazol
6-Fluoro-2-mercaptobenzothiazole
Formel: C₇H₄FNS₂
Molecular Weight: 185,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665747
CAS-Nummer: 80087-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR