Ethyl [3,5-bis(trifluoromethyl)benzoyl]acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 3-[3,5-bis(trifluoromethyl)phenyl]-3-oxopropanoate

PC32700-10G PC32700-1G
APOSPC32700-10GEA 0 EUR
APOSPC32700-10G APOSPC32700-1G
Ethyl [3,5-bis(trifluoromethyl)benzoyl]acetate
Ethyl [3,5-bis(trifluoromethyl)benzoyl]acetate
Formel: C₁₃H₁₀F₆O₃
Molecular Weight: 328,21 g/mol
Siedepunkt: 80…82 °C/0,6mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00052321
CAS-Nummer: 175278-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR