5-(Chlorosulphonyl)-2-fluorobenzoic acid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC3271-1G
PC3271-5G
PC3271-25G
APOSPC3271-1GEA
21.9
EUR
APOSPC3271-1G
APOSPC3271-5G
APOSPC3271-25G
5-(Chlorosulphonyl)-2-fluorobenzoic acid 98%
5-(Chlorosulphonyl)-2-fluorobenzoic acid
Formel:
C₇H₄ClFO₄S Molecular Weight: 238.613 g/mol Schmelzpunkt: 111…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03030335 CAS-Nummer: 37098-75-2 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...