2,2,2,2'-Tetrafluoracetophenon 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,2,2-Trifluoro-1-(2-fluorophenyl)ethanone
PC32719-1G
PC32719-250MG
PC32719-5G
APOSPC32719-1GEA
63.8
EUR
APOSPC32719-1G
APOSPC32719-250MG
APOSPC32719-5G
2,2,2,2'-Tetrafluoracetophenon 95%
2,2,2,2'-Tetrafluoracetophenon
Formel:
C₈H₄F₄O Molecular Weight: 192,1104 g/mol Siedepunkt: 60…62 °C/22mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040099 CAS-Nummer: 124004-75-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...