4-(Difluoromethyl)benzaldehyde

Lieferant: Apollo Scientific
Achtung

PC32743-1G PC32743-250MG
APOSPC32743-1GEA 415 EUR
APOSPC32743-1G APOSPC32743-250MG
4-(Difluoromethyl)benzaldehyde
4-(Difluoromethyl)benzaldehyde
Formel: C₈H₆F₂O
Molecular Weight: 156.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18072541
CAS-Nummer: 55805-29-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR