4-Brom-2-chlor-6-fluoriodbenzol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Bromo-1-chloro-3-fluoro-2-iodobenzene
PC32744-1G
PC32744-5G
PC32744-25G
APOSPC32744-1GEA
21.9
EUR
APOSPC32744-1G
APOSPC32744-5G
APOSPC32744-25G
4-Brom-2-chlor-6-fluoriodbenzol
4-Brom-2-chlor-6-fluoriodbenzol
Formel:
C₆H₂BrClFI Molecular Weight: 335,3400 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878165 CAS-Nummer: 83027-73-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...