3-Fluor-2-formylphenylboronsäure
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(3-Fluoro-2-formylphenyl)boronic acid, 3-Fluoro-2-formylbenzeneboronic acid
PC32758-1G
PC32758-25G
PC32758-5G
APOSPC32758-1GEA
21.9
EUR
APOSPC32758-1G
APOSPC32758-25G
APOSPC32758-5G
3-Fluor-2-formylphenylboronsäure
3-Fluor-2-formylphenylboronsäure
Formel:
C₇H₆BFO₃ Molecular Weight: 167.93 g/mol Schmelzpunkt: 123…127 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD10697421 CAS-Nummer: 871126-15-7 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...