2'-Brom-5'-fluoracetophenon

Lieferant: Apollo Scientific
Achtung

PC32800-10G PC32800-1G PC32800-5G
APOSPC32800-10GEA 36.4 EUR
APOSPC32800-10G APOSPC32800-1G APOSPC32800-5G
2'-Brom-5'-fluoracetophenon
2'-Brom-5'-fluoracetophenon
Formel: C₈H₆BrFO
Molecular Weight: 217.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07780635
CAS-Nummer: 1006-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR