Poly(chlortrifluorethylen) 800
Lieferant: Apollo Scientific
|
Achtung
|
PC32820-100G
PC32820-25G
APOSPC32820-100GEA
197
EUR
APOSPC32820-100G
APOSPC32820-25G
Poly(chlortrifluorethylen) 800
Poly(chlortrifluorethylen) 800
Inert, non-flammable lubricant for low temperature applications
Formel:
(C₂ClF₃)n Molecular Weight: 800 g/mol Siedepunkt: 132 °C Schmelzpunkt: < –130 °C Dichte: 1,71 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
CAS-Nummer:
9002-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...