6-Chloro-5-fluoro-1H-benzimidazol-2-amine

Lieferant: Apollo Scientific

PC32852-1G PC32852-50MG PC32852-250MG PC32852-500MG
APOSPC32852-1GEA 635 EUR
APOSPC32852-1G APOSPC32852-50MG APOSPC32852-250MG APOSPC32852-500MG
6-Chloro-5-fluoro-1H-benzimidazol-2-amine
6-Chloro-5-fluoro-1H-benzimidazol-2-amine
Formel: C₇H₅ClFN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18089180
CAS-Nummer: 142356-64-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR