5-Chloro-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

Lieferant: Apollo Scientific

PC32853-1G PC32853-250MG PC32853-500MG
APOSPC32853-1GEA 1110 EUR
APOSPC32853-1G APOSPC32853-250MG APOSPC32853-500MG
5-Chloro-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
5-Chloro-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Formel: C₅H₂ClF₃N₂O
Molecular Weight: 198.53 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07375187
CAS-Nummer: 154357-44-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR