N-(2-Bromo-6-fluorobenzyl)-N-methylamine

Lieferant: Apollo Scientific

PC32855-1G PC32855-250MG PC32855-500MG
APOSPC32855-1GEA 393 EUR
APOSPC32855-1G APOSPC32855-250MG APOSPC32855-500MG
N-(2-Bromo-6-fluorobenzyl)-N-methylamine
N-(2-Bromo-6-fluorobenzyl)-N-methylamine
Formel: C₈H₉BrFN
Lagertemperatur: Raumtemperatur
MDL: MFCD18089185
CAS-Nummer: 1287218-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR