Difluoro(6-methylpyridin-2-yl)acetic acid

Lieferant: Apollo Scientific

PC32862-250MG PC32862-500MG
APOSPC32862-250MGEA 328 EUR
APOSPC32862-250MG APOSPC32862-500MG
Difluoro(6-methylpyridin-2-yl)acetic acid
Difluoro(6-methylpyridin-2-yl)acetic acid
Formel: C₈H₇F₂NO₂
Molecular Weight: 187.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12827869
CAS-Nummer: 1215353-13-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR