3-(3-Fluorophenyl)acryloyl chloride

Lieferant: Apollo Scientific

PC32865-1G PC32865-250MG PC32865-500MG
APOSPC32865-1GEA 484 EUR
APOSPC32865-1G APOSPC32865-250MG APOSPC32865-500MG
3-(3-Fluorophenyl)acryloyl chloride
3-(3-Fluorophenyl)acryloyl chloride
Formel: C₉H₆ClFO
Lagertemperatur: Raumtemperatur
MDL: MFCD03424739
CAS-Nummer: 39098-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR