3-(4-Fluorophenyl)pyrazole
Lieferant: Apollo Scientific
|
Gefahr
|
PC3297-1G
PC3297-5G
PC3297-25G
APOSPC3297-1GEA
21.9
EUR
APOSPC3297-1G
APOSPC3297-5G
APOSPC3297-25G
3-(4-Fluorophenyl)pyrazole
3-(4-Fluorophenyl)pyrazole
Formel:
C₉H₇FN₂ Schmelzpunkt: 101…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00665857 CAS-Nummer: 154258-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...