2-(Trifluormethyl)-5-pyridinboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-(Trifluoromethyl)-3-pyridylboronic acid, 2-(Trifluoromethyl)pyridin-5-ylboronic acid, 2-(Trifluoromethyl)pyridine-5-boronic acid, (6-(Trifluoromethyl)pyridin-3-yl)boronic acid
PC3307-1G
PC3307-5G
APOSPC3307-1GEA
24.8
EUR
APOSPC3307-1G
APOSPC3307-5G
2-(Trifluormethyl)-5-pyridinboronsäure 97%
2-(Trifluormethyl)-5-pyridinboronsäure
Formel:
C₆H₅BF₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07375381 CAS-Nummer: 868662-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...