1-(4-Fluorphenylsulfonyl)pyrrolidin
Lieferant: Apollo Scientific
PC3357-1G
PC3357-5G
APOSPC3357-1GEA
72.7
EUR
APOSPC3357-1G
APOSPC3357-5G
1-(4-Fluorphenylsulfonyl)pyrrolidin
1-(4-Fluorphenylsulfonyl)pyrrolidin
Formel:
C₁₀H₁₂FNO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00783756 CAS-Nummer: 157187-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...