3'-Fluor-4'-(trifluormethyl)propiophenon 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC3366-250MG
PC3366-1G
PC3366-5G
APOSPC3366-250MGEA
21.9
EUR
APOSPC3366-250MG
APOSPC3366-1G
APOSPC3366-5G
3'-Fluor-4'-(trifluormethyl)propiophenon 97%
3'-Fluor-4'-(trifluormethyl)propiophenon
Formel:
C₁₀H₈F₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236306 CAS-Nummer: 237761-78-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...