3-Fluor-2-nitropyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC3369-1G
PC3369-5G
PC3369-10G
PC3369-25G
APOSPC3369-1GEA
21.9
EUR
APOSPC3369-1G
APOSPC3369-5G
APOSPC3369-10G
APOSPC3369-25G
3-Fluor-2-nitropyridin 98%
3-Fluoro-2-nitropyridine
Formel:
C₅H₃FN₂O₂ Schmelzpunkt: 30…35 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114127 CAS-Nummer: 54231-35-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...