3-Fluor-2-nitroanisol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC3402-1G
PC3402-5G
APOSPC3402-1GEA
21.9
EUR
APOSPC3402-1G
APOSPC3402-5G
3-Fluor-2-nitroanisol 98%
3-Fluor-2-nitroanisol
Formel:
C₇H₆FNO₃ Molecular Weight: 171,1259 g/mol Schmelzpunkt: 44…45 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368752 CAS-Nummer: 641-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...