3-Fluor-2-nitrophenol 98%

Lieferant: Apollo Scientific
Achtung

PC3403-5G PC3403-1G PC3403-25G
APOSPC3403-5GEA 24.8 EUR
APOSPC3403-5G APOSPC3403-1G APOSPC3403-25G
3-Fluor-2-nitrophenol 98%
3-Fluor-2-nitrophenol
Formel: C₆H₄FNO₃
Molecular Weight: 157,10 g/mol
Schmelzpunkt: 37…39 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368753
CAS-Nummer: 385-01-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR