2-Amino-3-fluorphenol 97%

Lieferant: Apollo Scientific
Achtung

PC3404-1G PC3404-5G
APOSPC3404-1GEA 23.2 EUR
APOSPC3404-1G APOSPC3404-5G
2-Amino-3-fluorphenol 97%
2-Amino-3-fluorphenol
Formel: C₆H₆FNO
Molecular Weight: 127.1163 g/mol
Schmelzpunkt: 126…127 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07368754
CAS-Nummer: 53981-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR