5-Fluor-2-(trifluormethyl)benzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
α,α,α,5-Tetrafluoro-o-tolualdehyde, 2-Trifluoromethyl-5-fluorobenzaldehyde
PC3423-10G
PC3423-5G
APOSPC3423-10GEA
79.9
EUR
APOSPC3423-10G
APOSPC3423-5G
5-Fluor-2-(trifluormethyl)benzaldehyd 98%
5-Fluor-2-(trifluormethyl)benzaldehyd
Formel:
C₈H₄F₄O Molecular Weight: 192,11 g/mol Flammpunkt: 82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236276 CAS-Nummer: 90381-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...