8-Fluorisochinolin
Lieferant: Apollo Scientific
|
Achtung
|
PC3441-100MG
PC3441-500MG
PC3441-250MG
APOSPC3441-100MGEA
21.9
EUR
APOSPC3441-100MG
APOSPC3441-500MG
APOSPC3441-250MG
8-Fluorisochinolin
8-Fluoroisoquinoline
Formel:
C₉H₆FN Schmelzpunkt: 27…28 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09878369 CAS-Nummer: 1075-00-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...