2-Fluorbenzalchlorid 97%
Lieferant: Apollo Scientific
Synonyme:
α,α-Dichloro-2-fluortoluene
PC3444A-25G
PC3444A-5G
PC3444A-100G
APOSPC3444A-25GEA
216
EUR
APOSPC3444A-25G
APOSPC3444A-5G
APOSPC3444A-100G
2-Fluorbenzalchlorid 97%
2-Fluorbenzalchlorid
Formel:
C₇H₅Cl₂F Molecular Weight: 179,02 g/mol Siedepunkt: 224…232 °C Dichte: 1,309 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043797 CAS-Nummer: 320-65-0 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...