2,3,5-Trifluorbenzamid 97%

Lieferant: Apollo Scientific
Achtung

PC3445-5G PC3445-1G
APOSPC3445-5GEA 59.6 EUR
APOSPC3445-5G APOSPC3445-1G
2,3,5-Trifluorbenzamid 97%
2,3,5-Trifluorbenzamid
Formel: C₇H₄F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00061198
CAS-Nummer: 238403-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR