4-Fluorphenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Dihydroxy(4-fluorphenyl)boran, 4-Fluorbenzolboronsäure, (4-Fluorphenyl)metaborsäure
PC3482-25G
PC3482-100G
PC3482-500G
APOSPC3482-25GEA
21.9
EUR
APOSPC3482-25G
APOSPC3482-100G
APOSPC3482-500G
4-Fluorphenylboronsäure 97%
4-Fluorphenylboronsäure
Contains variable amounts of the anhydride GLC reagent for diols see: J. Chromatogr.,158,33,1978
Formel:
C₆H₆BFO₂ Molecular Weight: 139,92 g/mol Schmelzpunkt: 262…265 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039136 CAS-Nummer: 1765-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...